Assay Details
Target Gene Details
Entrez Gene ID: | 9601 |
Gene Name: | protein disulfide isomerase family A member 4 |
Gene Aliases: |
ERP70, ERP72, ERp-72 |
Location: |
Chr.7:149003054-149028690 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDIA4 | NM_004911.4 | 10 | 2369 | NP_004902.1 |
| XM_006716185.2 | 10 | 2372 | XP_006716248.1 | |
| AK290971.1 | 10 | 2154 | ||
| BC000425.2 | 10 | 2131 | AAH00425.1 | |
| BC001928.1 | 10 | 2131 | AAH01928.1 | |
| BC006344.2 | 10 | 2319 | AAH06344.1 | |
| BC011754.2 | 10 | 2320 | AAH11754.1 | |
| BX330378.1 | 1 | 201 | ||
| CX871856.1 | 1 | 224 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528358 | Chr.7:148964410 - 149040285 on Build GRCh38 | Loss |
|
| nsv608973 | Chr.7:148976273 - 149069299 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map