Assay Details
Target Gene Details
Entrez Gene ID: | 8633 |
Gene Name: | unc-5 netrin receptor C |
Gene Aliases: |
UNC5H3 |
Location: |
Chr.4:95162504-95549210 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC5C | NM_003728.3 | 16 | 3793 | NP_003719.3 |
| XM_005263321.3 | 17 | 3850 | XP_005263378.1 | |
| XM_017008789.1 | 17 | 10210 | XP_016864278.1 | |
| XM_017008790.1 | 16 | 3438 | XP_016864279.1 | |
| AF055634.1 | 16 | 3594 | AAC67491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830011 | Chr.4:95026409 - 95182783 on Build GRCh38 | Gain |
|
| nsv594907 | Chr.4:95054319 - 95249288 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map