Assay Details
Target Gene Details
Entrez Gene ID: | 6782 |
Gene Name: | heat shock protein family A (Hsp70) member 13 |
Gene Aliases: |
STCH |
Location: |
Chr.21:14371115-14383188 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPA13 | NM_006948.4 | 5 | 1446 | NP_008879.3 |
| AK128540.1 | 5 | 1228 | ||
| AK223282.1 | 5 | 1404 | BAD97002.1 | |
| AK299004.1 | 4 | 1181 | ||
| AK312396.1 | ||||
| BC036370.2 | 5 | 1408 | AAH36370.1 | |
| U04735.1 | 5 | 1413 | AAA16954.1 |
More Information
Set Membership: |
|

Genomic Map