Assay Details
Target Gene Details
Entrez Gene ID: | 90627 |
Gene Name: | StAR related lipid transfer domain containing 13 |
Gene Aliases: |
ARHGAP37, DLC2, GT650, LINC00464 |
Location: |
Chr.13:33103135-33676835 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD13 | NM_001243476.2 | 18 | 5995 | NP_001230405.1 |
| NM_052851.2 | 14 | 5585 | NP_443083.1 | |
| NM_178006.3 | 14 | 5648 | NP_821074.1 | |
| NM_178007.2 | 14 | 5555 | NP_821075.1 | |
| XM_011535299.2 | 17 | 5741 | XP_011533601.1 | |
| XM_017020834.1 | 14 | 5753 | XP_016876323.1 | |
| XM_017020835.1 | 16 | 5674 | XP_016876324.1 | |
| AA256386.1 | 1 | 282 | ||
| AL049801.1 | 14 | 5541 | CAB42562.1 | |
| AY082589.1 | 14 | 5643 | AAL91648.1 | |
| AY082590.1 | 14 | 5567 | AAL91649.1 | |
| AY082591.1 | 14 | 5541 | AAL91650.1 | |
| BX641149.1 | 4 | 2668 | CAE46065.1 | |
| BX647695.1 | 18 | 5955 | CAI46026.1 |
More Information
Set Membership: |
|

Genomic Map