Assay Details
Target Gene Details
Entrez Gene ID: | 54795 |
Gene Name: | transient receptor potential cation channel subfamily M member 4 |
Gene Aliases: |
LTrpC4, PFHB1B, TRPM4B, hTRPM4 |
Location: |
Chr.19:49157741-49211841 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPM4 | NM_001195227.1 | NP_001182156.1 | ||
| NM_001321281.1 | NP_001308210.1 | |||
| NM_001321282.1 | NP_001308211.1 | |||
| NM_001321283.1 | NP_001308212.1 | |||
| NM_001321285.1 | NP_001308214.1 | |||
| NM_017636.3 | NP_060106.2 | |||
| AF497623.1 | AAM18083.1 | |||
| AJ575813.1 | CAE05941.1 | |||
| AK000048.1 | BAA90907.1 | |||
| AK292862.1 | ||||
| AK295820.1 | ||||
| AK307897.1 | ||||
| AY046396.1 | AAL02142.1 | |||
| AY297044.1 | AAP44473.1 | |||
| AY297045.1 | AAP44474.1 | |||
| AY297046.1 | AAP44475.1 | |||
| BC132727.1 | ||||
| DA218254.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833859 | Chr.19:48997009 - 49173266 on Build GRCh38 | Loss |
|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv1060108 | Chr.19:49118098 - 49183334 on Build GRCh38 | Gain |
|
| dgv1057e212 | Chr.19:49165806 - 49183334 on Build GRCh38 | Loss |
|
| esv2760526 | Chr.19:49115521 - 49170761 on Build GRCh38 | Gain |
|
| dgv667e214 | Chr.19:49130606 - 49174364 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111783027,rs115335683] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map