Assay Details
Target Gene Details
Entrez Gene ID: | 164045 |
Gene Name: | HFM1, ATP dependent DNA helicase homolog |
Gene Aliases: |
MER3, POF9, SEC63D1, Si-11, Si-11-6, helicase |
Location: |
Chr.1:91260766-91408008 on Build GRCh38 |
Assay Gene Location: | Within Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HFM1 | NM_001017975.4 | 35 | 3918 | NP_001017975.4 |
| XM_011540849.1 | 35 | 4001 | XP_011539151.1 | |
| XM_011540850.2 | 35 | 3930 | XP_011539152.1 | |
| XM_011540851.1 | 35 | 3885 | XP_011539153.1 | |
| XM_011540852.2 | 35 | 3867 | XP_011539154.1 | |
| XM_011540855.1 | 33 | 3723 | XP_011539157.1 | |
| XM_011540857.1 | 33 | 3482 | XP_011539159.1 | |
| XM_011540859.2 | 32 | 3451 | XP_011539161.1 | |
| XM_017000490.1 | 34 | 3781 | XP_016855979.1 | |
| XM_017000491.1 | 34 | 3730 | XP_016855980.1 | |
| XM_017000492.1 | 29 | 2965 | XP_016855981.1 | |
| XM_017000493.1 | 23 | 2169 | XP_016855982.1 | |
| AB044787.1 | 12 | 1206 | ||
| AB074274.1 | 7 | 669 | ||
| AB204867.1 | 35 | 3919 | ||
| AK094079.1 | 18 | 1745 | BAC04281.1 | |
| AK294754.1 | 29 | 2936 | ||
| AK302582.1 | 31 | 3086 | ||
| BC144172.1 | 20 | 1864 | ||
| BX649037.1 | 16 | 1454 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map