Assay Details
Target Gene Details
Entrez Gene ID: | 100129792 |
Gene Name: | coiled-coil domain containing 152 |
Gene Aliases: |
CH5400 |
Location: |
Chr.5:42756818-42802437 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC152 | NM_001134848.1 | 9 | 1608 | NP_001128320.1 |
| AK127368.1 | 9 | 1598 | ||
| BC039102.1 | 9 | 1580 | AAH39102.1 |
Target Gene Details
Entrez Gene ID: | 6414 |
Gene Name: | selenoprotein P, plasma, 1 |
Gene Aliases: |
SELP, SEPP, SeP |
Location: |
Chr.5:42799880-42811922 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPP1 | NM_001085486.1 | 6 | 1441 | NP_001078955.1 |
| NM_001093726.1 | 6 | 1500 | NP_001087195.1 | |
| NM_005410.2 | 5 | 1412 | NP_005401.3 | |
| AK094640.1 | 1 | 1445 | ||
| AK096125.1 | 5 | 1781 | ||
| AK225801.1 | 6 | 1411 | ||
| AL833145.1 | 5 | 1383 | ||
| BC015875.1 | 5 | 1386 | AAH15875.1 | |
| BC040075.1 | 5 | 1362 | AAH40075.1 | |
| BC046152.1 | 5 | 1363 | AAH46152.1 | |
| BC058919.1 | 5 | 1393 | AAH58919.1 | |
| Z11793.1 | 5 | 1348 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5637n100 | Chr.5:42609988 - 42844089 on Build GRCh38 | Gain |
|
| nsv597886 | Chr.5:42610307 - 42854306 on Build GRCh38 | Gain |
|
| nsv523033 | Chr.5:42751935 - 42919583 on Build GRCh38 | Gain |
|
| nsv462126 | Chr.5:42728729 - 43237086 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map