Assay Details
Target Gene Details
Entrez Gene ID: | 23657 |
Gene Name: | solute carrier family 7 member 11 |
Gene Aliases: |
CCBR1, xCT |
Location: |
Chr.4:138164094-138242418 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A11 | NM_014331.3 | 12 | 7340 | NP_055146.1 |
| AB040875.1 | BAB40574.1 |
Target Gene Details
Entrez Gene ID: | 641364 |
Gene Name: | SLC7A11 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:138089014-138178177 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A11-AS1 | NR_038380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830080 | Chr.4:138031664 - 138223572 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map