Assay Details
Target Gene Details
Entrez Gene ID: | 783 |
Gene Name: | calcium voltage-gated channel auxiliary subunit beta 2 |
Gene Aliases: |
CACNLB2, CAVB2, MYSB |
Location: |
Chr.10:18140560-18545881 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNB2 | NM_000724.3 | 13 | 2878 | NP_000715.2 |
| NM_001167945.1 | 14 | 2597 | NP_001161417.1 | |
| NM_201570.2 | 13 | 2732 | NP_963864.1 | |
| NM_201571.3 | 14 | 2711 | NP_963865.2 | |
| NM_201572.3 | 14 | 2639 | NP_963866.2 | |
| NM_201590.2 | 13 | 2508 | NP_963884.2 | |
| NM_201593.2 | 14 | 2374 | NP_963887.2 | |
| NM_201596.2 | 14 | 2488 | NP_963890.2 | |
| NM_201597.2 | 14 | 2416 | NP_963891.1 | |
| XM_005252588.3 | 13 | 2622 | XP_005252645.1 | |
| XM_005252591.3 | 8 | 1683 | XP_005252648.1 | |
| XM_006717502.3 | 13 | 2541 | XP_006717565.1 | |
| XM_011519659.2 | 13 | 2436 | XP_011517961.1 | |
| XM_011519660.2 | 13 | 2979 | XP_011517962.1 | |
| XM_017016625.1 | 14 | 2596 | XP_016872114.1 | |
| AB208917.1 | 13 | 2764 | BAD92154.1 | |
| S60415.1 | 14 | 2488 | AAB51370.1 | |
| U95019.1 | 13 | 2764 | AAB53332.1 |
Target Gene Details
Entrez Gene ID: | 221078 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 6 |
Gene Aliases: |
4933414E04Rik, ARL5B-AS1, NOPD1 |
Location: |
Chr.10:18512582-18659264 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSUN6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042073 | Chr.10:18517971 - 18549374 on Build GRCh38 | Gain |
|
| nsv550042 | Chr.10:18394338 - 18705654 on Build GRCh38 | Gain |
|
| esv34545 | Chr.10:18532256 - 18619365 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map