Assay Details
Target Gene Details
Entrez Gene ID: | 124995 |
Gene Name: | mitochondrial ribosomal protein L10 |
Gene Aliases: |
L10MT, MRP-L10, MRP-L8, MRPL8, RPML8 |
Location: |
Chr.17:47823272-47831541 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL10 | NM_145255.3 | NP_660298.2 | ||
| NM_148887.2 | NP_683685.1 | |||
| NR_037575.1 | ||||
| AK127167.1 | ||||
| AK293624.1 | ||||
| AL558270.3 | ||||
| AL701268.1 | ||||
| BC015904.1 | AAH15904.1 | |||
| BC052601.1 | AAH52601.1 | |||
| DA137043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv947e212 | Chr.17:47826629 - 47839274 on Build GRCh38 | Loss |
|
| nsv518892 | Chr.17:47810585 - 47952066 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map