Assay Details
Target Gene Details
Entrez Gene ID: | 10551 |
Gene Name: | anterior gradient 2, protein disulphide isomerase family member |
Gene Aliases: |
AG2, GOB-4, HAG-2, HEL-S-116, PDIA17, XAG-2 |
Location: |
Chr.7:16791811-16805114 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGR2 | XM_005249581.4 | 8 | 1054 | XP_005249638.1 |
| AF038451.1 | 8 | 956 | AAC82614.1 | |
| AF038452.1 | 1 | 113 | ||
| AF115926.1 | 8 | 955 | AAL54870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6289n100 | Chr.7:16230707 - 17452994 on Build GRCh38 | Gain |
|
| nsv1028125 | Chr.7:16489535 - 16807601 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map