Assay Details
Target Gene Details
Entrez Gene ID: | 124995 |
Gene Name: | mitochondrial ribosomal protein L10 |
Gene Aliases: |
L10MT, MRP-L10, MRP-L8, MRPL8, RPML8 |
Location: |
Chr.17:47823272-47831541 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL10 | NM_145255.3 | 5 | 844 | NP_660298.2 |
| NM_148887.2 | 6 | 942 | NP_683685.1 | |
| NR_037575.1 | 5 | 993 | ||
| AK127167.1 | 5 | 1719 | ||
| AK293624.1 | 5 | 982 | ||
| AL558270.3 | 6 | 907 | ||
| BC015904.1 | 5 | 815 | AAH15904.1 | |
| BC052601.1 | 5 | 833 | AAH52601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518892 | Chr.17:47810585 - 47952066 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map