Assay Details
Target Gene Details
Entrez Gene ID: | 54476 |
Gene Name: | ring finger protein 216 |
Gene Aliases: |
CAHH, TRIAD3, U7I1, UBCE7IP1, ZIN |
Location: |
Chr.7:5620041-5781730 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF216 | NM_207111.3 | NP_996994.1 | ||
| NM_207116.2 | NP_996999.1 | |||
| XM_005249785.2 | XP_005249842.1 | |||
| XM_011515436.1 | XP_011513738.1 | |||
| XM_017012363.1 | XP_016867852.1 | |||
| XM_017012365.1 | XP_016867854.1 | |||
| BM970811.1 | ||||
| BM997118.1 | ||||
| BQ018569.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830890 | Chr.7:5425302 - 5665527 on Build GRCh38 | Loss |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
| nsv1034883 | Chr.7:5612049 - 5656516 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map