Assay Details
Target Gene Details
Entrez Gene ID: | 84947 |
Gene Name: | serine active site containing 1 |
Gene Aliases: |
- |
Location: |
Chr.6:158109504-158168280 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERAC1 | NM_032861.3 | 17 | 2233 | NP_116250.3 |
| XM_006715586.2 | 16 | 2162 | XP_006715649.1 | |
| XM_011536196.2 | 16 | 2256 | XP_011534498.1 | |
| XM_011536198.2 | 15 | 2158 | XP_011534500.1 | |
| AK023656.1 | 1 | 269 | ||
| AK027823.1 | 17 | 2223 | BAB55393.1 | |
| AK125878.1 | 18 | 3425 | ||
| BC001705.1 | 17 | 2190 | AAH01705.1 | |
| BX648104.1 | 15 | 3609 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890972 | Chr.6:158082895 - 158303958 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map