Assay Details
Target Gene Details
Entrez Gene ID: | 9547 |
Gene Name: | C-X-C motif chemokine ligand 14 |
Gene Aliases: |
BMAC, BRAK, KEC, KS1, MIP-2g, MIP2G, NJAC, SCYB14 |
Location: |
Chr.5:135570679-135579279 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL14 | NM_004887.4 | 4 | 1738 | NP_004878.2 |
| AF144103.1 | 4 | 1244 | AAD38944.1 | |
| AK075514.1 | 4 | 1455 | ||
| AY358906.1 | 4 | 1439 | AAQ89265.1 | |
| BM670989.1 | 1 | 249 | ||
| BU729234.1 | 1 | 252 |
More Information
Set Membership: |
|

Genomic Map