Assay Details
Target Gene Details
Entrez Gene ID: | 56897 |
Gene Name: | Werner helicase interacting protein 1 |
Gene Aliases: |
WHIP, bA420G6.2 |
Location: |
Chr.6:2765341-2785745 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WRNIP1 | NM_020135.2 | 7 | 2276 | NP_064520.2 |
| NM_130395.2 | 7 | 2201 | NP_569079.1 | |
| AB056152.1 | 7 | 2277 | BAB60709.1 | |
| AB209723.1 | 7 | 2029 | BAD92960.1 | |
| AF218313.1 | 7 | 1663 | AAF80563.1 | |
| AK026179.1 | 6 | 1181 | BAB15383.1 | |
| AK094823.1 | 6 | 2083 | ||
| AK223593.1 | BAD97313.1 | |||
| BC018923.2 | 7 | 2162 | AAH18923.1 |
More Information
Set Membership: |
|

Genomic Map