Assay Details
Target Gene Details
Entrez Gene ID: | 3689 |
Gene Name: | integrin subunit beta 2 |
Gene Aliases: |
CD18, LAD, LCAMB, LFA-1, MAC-1, MF17, MFI7 |
Location: |
Chr.21:44885949-44928873 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB2 | NM_000211.4 | 15 | 2424 | NP_000202.3 |
| NM_001127491.2 | 15 | 2414 | NP_001120963.2 | |
| NM_001303238.1 | 15 | 2379 | NP_001290167.1 | |
| XM_006724001.1 | 15 | 2441 | XP_006724064.1 | |
| XM_017028341.1 | 15 | 2436 | XP_016883830.1 | |
| AB208909.1 | 14 | 4247 | BAD92146.1 | |
| AK095992.1 | 15 | 2379 | ||
| AK097216.1 | 6 | 1591 | ||
| AK097864.1 | 1 | 1825 | ||
| AK128344.1 | 15 | 2249 | ||
| AK222505.1 | 15 | 2281 | BAD96225.1 | |
| AK303488.1 | 15 | 2292 | ||
| BC005861.2 | 14 | 2235 | AAH05861.1 | |
| BC021077.1 | 14 | 2158 | AAH21077.1 | |
| CA447686.1 | 2 | 550 | ||
| L78790.1 | 14 | 2071 | AAL40068.1 | |
| M15395.1 | 15 | 2263 | AAA59490.1 | |
| Y00057.1 | 14 | 2167 | CAA68266.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv459301 | Chr.21:44886246 - 44902938 on Build GRCh38 | Loss |
|
| nsv1058550 | Chr.21:44807248 - 44898019 on Build GRCh38 | Loss |
|
| esv3647136 | Chr.21:44802580 - 44905193 on Build GRCh38 | Loss |
|
| nsv587814 | Chr.21:44863754 - 44891349 on Build GRCh38 | Gain |
|
| nsv470908 | Chr.21:44878954 - 44915665 on Build GRCh38 | Loss |
|
| nsv953648 | Chr.21:44883286 - 44909585 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map