Assay Details
Target Gene Details
Entrez Gene ID: | 8925 |
Gene Name: | HECT and RLD domain containing E3 ubiquitin protein ligase family member 1 |
Gene Aliases: |
p532, p619 |
Location: |
Chr.15:63608618-63833964 on Build GRCh38 |
Assay Gene Location: | Within Exon 69 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HERC1 | NM_003922.3 | 67 | 12780 | NP_003913.3 |
| XM_017022699.1 | 68 | 12937 | XP_016878188.1 | |
| XM_017022700.1 | 68 | 12901 | XP_016878189.1 | |
| XM_017022701.1 | 68 | 12886 | XP_016878190.1 | |
| XM_017022702.1 | 67 | 12886 | XP_016878191.1 | |
| XM_017022703.1 | 68 | 12883 | XP_016878192.1 | |
| XM_017022704.1 | 68 | 12865 | XP_016878193.1 | |
| XM_017022705.1 | 67 | 12859 | XP_016878194.1 | |
| AK226162.1 | 54 | 9843 | ||
| BC040929.1 | AAH40929.1 | |||
| U50078.1 | 67 | 12728 | AAD12586.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833034 | Chr.15:63500671 - 63657512 on Build GRCh38 | Loss |
|
| nsv833035 | Chr.15:63633089 - 63807741 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map