Assay Details
Target Gene Details
Entrez Gene ID: | 84749 |
Gene Name: | ubiquitin specific peptidase 30 |
Gene Aliases: |
- |
Location: |
Chr.12:109023089-109088026 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP30 | NM_001301175.1 | 6 | 749 | NP_001288104.1 |
| NM_032663.4 | 3 | 375 | NP_116052.2 | |
| XM_005253962.3 | 3 | 361 | XP_005254019.1 | |
| XM_005253965.4 | XP_005254022.1 | |||
| XM_006719653.3 | 4 | 448 | XP_006719716.1 | |
| XM_011538894.2 | XP_011537196.1 | |||
| XM_017020048.1 | 3 | 370 | XP_016875537.1 | |
| XM_017020049.1 | 3 | 361 | XP_016875538.1 | |
| XM_017020050.1 | 3 | 470 | XP_016875539.1 | |
| XM_017020051.1 | 4 | 326 | XP_016875540.1 | |
| XM_017020052.1 | 3 | 372 | XP_016875541.1 | |
| XM_017020053.1 | XP_016875542.1 | |||
| XM_017020054.1 | XP_016875543.1 | |||
| AJ586136.1 | 3 | 266 | CAE51936.1 | |
| AK027820.1 | 3 | 340 | BAB55392.1 | |
| AK097830.1 | 6 | 749 | ||
| BC004868.2 | 3 | 365 | AAH04868.1 | |
| HY127211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422361 | Chr.12:108792678 - 109118360 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map