Assay Details
Target Gene Details
Entrez Gene ID: | 23284 |
Gene Name: | adhesion G protein-coupled receptor L3 |
Gene Aliases: |
CIRL3, CL3, LEC3, LPHN3 |
Location: |
Chr.4:61201256-62078335 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRL3 | NM_001322246.1 | 26 | 6308 | NP_001309175.1 |
| NM_001322402.1 | 26 | 6460 | NP_001309331.1 | |
| NM_015236.5 | 25 | 6256 | NP_056051.2 | |
| XM_011531788.2 | 27 | 6016 | XP_011530090.1 | |
| XM_011531791.2 | 24 | 5932 | XP_011530093.1 | |
| XM_017007929.1 | 29 | 8918 | XP_016863418.1 | |
| XM_017007930.1 | 28 | 7870 | XP_016863419.1 | |
| XM_017007931.1 | 27 | 6127 | XP_016863420.1 | |
| XM_017007932.1 | 26 | 5998 | XP_016863421.1 | |
| XM_017007933.1 | 26 | 5989 | XP_016863422.1 | |
| XM_017007934.1 | 26 | 5983 | XP_016863423.1 | |
| XM_017007935.1 | 25 | 5971 | XP_016863424.1 | |
| XM_017007936.1 | 25 | 5959 | XP_016863425.1 | |
| XM_017007937.1 | 27 | 7666 | XP_016863426.1 | |
| XM_017007938.1 | 24 | 5917 | XP_016863427.1 | |
| XM_017007939.1 | 22 | 5713 | XP_016863428.1 | |
| XM_017007940.1 | 27 | 6050 | XP_016863429.1 | |
| XM_017007941.1 | 26 | 6023 | XP_016863430.1 | |
| XM_017007942.1 | 26 | 5831 | XP_016863431.1 | |
| AB018311.1 | 13 | 3717 | BAA34488.1 | |
| AK000781.1 | 7 | 1499 | BAA91375.1 |
Target Gene Details
Entrez Gene ID: | 101927186 |
Gene Name: | adhesion G protein-coupled receptor L3 antisense RNA 1 |
Gene Aliases: |
LPHN3-AS1 |
Location: |
Chr.4:62071755-62161765 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRL3-AS1 | NR_110595.1 | |||
| DB518756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012574 | Chr.4:61978546 - 62088469 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map