Assay Details
Target Gene Details
Entrez Gene ID: | 27190 |
Gene Name: | interleukin 17B |
Gene Aliases: |
IL-17B, IL-20, NIRF, ZCYTO7 |
Location: |
Chr.5:149374267-149404202 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL17B | NM_001317987.1 | NP_001304916.1 | ||
| NM_014443.2 | NP_055258.1 | |||
| XM_011537622.2 | XP_011535924.1 | |||
| XM_017009346.1 | XP_016864835.1 | |||
| XM_017009347.1 | XP_016864836.1 | |||
| XM_017009348.1 | XP_016864837.1 | |||
| AF110385.1 | AAG39637.1 | |||
| AF152098.1 | AAF28104.1 | |||
| AF184969.1 | AAF01318.1 | |||
| AF212311.1 | AAF78775.1 | |||
| AF218727.1 | AAG44136.1 | |||
| AY358931.1 | AAQ89290.1 | |||
| BC113925.1 | ||||
| BC113946.1 | ||||
| BM826445.1 | ||||
| CR457192.1 | CAG33473.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599948 | Chr.5:149346599 - 149413198 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map