Assay Details
Target Gene Details
Entrez Gene ID: | 64975 |
Gene Name: | mitochondrial ribosomal protein L41 |
Gene Aliases: |
BMRP, MRP-L27, MRPL27, PIG3, RPML27 |
Location: |
Chr.9:137551857-137552555 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL41 | NM_032477.2 | 2 | 346 | NP_115866.1 |
| AI091177.1 | 2 | 346 | ||
| AY232291.1 | 2 | 270 | AAP69986.1 | |
| BC040035.1 | 2 | 321 | AAH40035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831762 | Chr.9:137444327 - 137619655 on Build GRCh38 | Loss |
|
| nsv616023 | Chr.9:137478118 - 137708435 on Build GRCh38 | Loss |
|
| nsv831763 | Chr.9:137545379 - 137719569 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map