Assay Details
Target Gene Details
Entrez Gene ID: | 9249 |
Gene Name: | dehydrogenase/reductase 3 |
Gene Aliases: |
DD83.1, RDH17, Rsdr1, SDR1, SDR16C1, retSDR1 |
Location: |
Chr.1:12567910-12618361 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHRS3 | NM_001319225.1 | NP_001306154.1 | ||
| NM_001324370.1 | NP_001311299.1 | |||
| NM_004753.6 | NP_004744.2 | |||
| XM_006711036.1 | XP_006711099.1 | |||
| XM_017002809.1 | XP_016858298.1 | |||
| AF061741.1 | AAC63263.1 | |||
| AK222593.1 | BAD96313.1 | |||
| AK225057.1 | ||||
| AK312961.1 | ||||
| BC002730.2 | AAH02730.1 | |||
| BX648476.1 | CAI46033.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517848 | Chr.1:12313691 - 12582215 on Build GRCh38 | Gain |
|
| nsv521737 | Chr.1:12539396 - 12624831 on Build GRCh38 | Loss |
|
| nsv545442 | Chr.1:12565072 - 12610391 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map