Assay Details
Target Gene Details
Entrez Gene ID: | 60386 |
Gene Name: | solute carrier family 25 member 19 |
Gene Aliases: |
DNC, MCPHA, MUP1, THMD3, THMD4, TPC |
Location: |
Chr.17:75272980-75289957 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A19 | NM_001126121.1 | 3 | 285 | NP_001119593.1 |
| NM_001126122.1 | 2 | 195 | NP_001119594.1 | |
| NM_021734.4 | 3 | 269 | NP_068380.3 | |
| XM_005257559.3 | 3 | 907 | XP_005257616.1 | |
| XM_005257560.2 | 3 | 461 | XP_005257617.1 | |
| XM_005257561.3 | 3 | 797 | XP_005257618.1 | |
| XM_005257562.2 | 3 | 444 | XP_005257619.1 | |
| XM_006722007.2 | 3 | 317 | XP_006722070.1 | |
| XM_011525098.1 | 3 | 777 | XP_011523400.1 | |
| XM_017024926.1 | 2 | 859 | XP_016880415.1 | |
| XM_017024927.1 | XP_016880416.1 | |||
| XM_017024928.1 | 3 | 793 | XP_016880417.1 | |
| AF182404.1 | 1 | 61 | AAG16903.1 | |
| AJ251857.1 | 1 | 99 | CAC27560.1 | |
| AK075239.1 | 4 | 291 | BAC11492.1 | |
| AK097882.1 | 1 | 971 | ||
| AK225881.1 | 1 | 98 | ||
| AL833387.1 | 3 | 236 | CAI46136.1 | |
| AY346372.1 | 1 | 61 | AAQ54327.1 | |
| BC001075.2 | 3 | 197 | AAH01075.1 | |
| BC005120.1 | 3 | 197 | AAH05120.1 | |
| BM455504.1 | ||||
| BU177306.1 | 2 | 154 | ||
| DA605220.1 | 3 | 250 | ||
| DB482052.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510727 | Chr.17:75260216 - 75331456 on Build GRCh38 | Deletion |
|
| nsv1066312 | Chr.17:75260266 - 75323694 on Build GRCh38 | Gain |
|
| nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs546901072] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map