Assay Details
Target Gene Details
Entrez Gene ID: | 55615 |
Gene Name: | proline rich 5 |
Gene Aliases: |
FLJ20185k, PP610, PROTOR-1, PROTOR1 |
Location: |
Chr.22:44668547-44737681 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRR5 | NM_001017528.2 | 9 | 1079 | NP_001017528.1 |
| NM_001017529.2 | 8 | 962 | NP_001017529.1 | |
| NM_001017530.1 | 8 | 1136 | NP_001017530.1 | |
| NM_001198721.1 | 10 | 1792 | NP_001185650.1 | |
| NM_015366.3 | 9 | 1253 | NP_056181.2 | |
| NM_181333.3 | 8 | 1114 | NP_851850.1 | |
| AF177331.1 | 8 | 1058 | AAG17975.1 | |
| AK000192.1 | 4 | 1172 | BAA90999.1 | |
| AK055848.1 | 10 | 1515 | ||
| AK056787.1 | 2 | 1200 | ||
| AK091884.1 | 9 | 1004 | ||
| AK291564.1 | 8 | 1104 | ||
| BC016921.1 | 4 | 1361 | AAH16921.1 |
Target Gene Details
Entrez Gene ID: | 553158 |
Gene Name: | PRR5-ARHGAP8 readthrough |
Gene Aliases: |
- |
Location: |
Chr.22:44702198-44862784 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRR5-ARHGAP8 | NM_181334.5 | NP_851851.3 | ||
| AF195968.1 | AAK58136.1 | |||
| AF195969.1 | AAK58137.1 | |||
| AL355192.1 | CAB90248.1 | |||
| BC132755.1 | ||||
| BC132757.1 | ||||
| BC144382.1 | ||||
| BI488517.1 | ||||
| BX335168.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1325e212 | Chr.22:44734370 - 44741324 on Build GRCh38 | Loss |
|
| nsv589108 | Chr.22:44721714 - 44796741 on Build GRCh38 | Loss |
|
| nsv589109 | Chr.22:44725984 - 44739384 on Build GRCh38 | Loss |
|
| nsv829268 | Chr.22:44640618 - 44785937 on Build GRCh38 | Gain |
|
| nsv817999 | Chr.22:44698163 - 44737460 on Build GRCh38 | Gain |
|
| esv2758842 | Chr.22:44630924 - 44814649 on Build GRCh38 | Loss |
|
| dgv4586n100 | Chr.22:44719617 - 44799761 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map