Assay Details
Target Gene Details
Entrez Gene ID: | 11269 |
Gene Name: | DEAD-box helicase 19B |
Gene Aliases: |
DBP5, DDX19, RNAh |
Location: |
Chr.16:70289767-70335213 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Target Gene Details
Entrez Gene ID: | 100506083 |
Gene Name: | uncharacterized LOC100506083 |
Gene Aliases: |
- |
Location: |
Chr.16:70315640-70346747 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506083 | NR_039997.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74024199] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map