Assay Details
Target Gene Details
Entrez Gene ID: | 57709 |
Gene Name: | solute carrier family 7 member 14 |
Gene Aliases: |
PPP1R142 |
Location: |
Chr.3:170459554-170586074 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A14 | NM_020949.2 | 8 | 2472 | NP_066000.2 |
| AB046833.1 | 7 | 1923 | BAB13439.1 | |
| AK122655.1 | 8 | 2467 | ||
| BC022968.1 | 7 | 2088 |
More Information
Additional Information:
For this assay, SNP(s) [rs73040363] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map