Assay Details
Target Gene Details
Entrez Gene ID: | 200150 |
Gene Name: | phospholipase D family member 5 |
Gene Aliases: |
PLDC |
Location: |
Chr.1:242085051-242530546 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLD5 | NM_001195811.1 | 10 | 1905 | NP_001182740.1 |
| NM_001195812.1 | 9 | 1743 | NP_001182741.1 | |
| NM_001320272.1 | 11 | 2199 | NP_001307201.1 | |
| NM_152666.2 | 11 | 2192 | NP_689879.2 | |
| XM_006711752.2 | 10 | 2359 | XP_006711815.1 | |
| XM_011544115.2 | 12 | 2267 | XP_011542417.1 | |
| XM_011544116.2 | 12 | 2095 | XP_011542418.1 | |
| XM_011544119.2 | 7 | 2266 | XP_011542421.2 | |
| XM_011544120.2 | 10 | 1911 | XP_011542422.1 | |
| XM_011544121.2 | 7 | 1400 | XP_011542423.1 | |
| XM_011544122.2 | 7 | 2861 | XP_011542424.1 | |
| XM_017000567.1 | 10 | 2372 | XP_016856056.1 | |
| XM_017000568.1 | 10 | 1893 | XP_016856057.1 | |
| XM_017000569.1 | 10 | 3872 | XP_016856058.1 | |
| XM_017000570.1 | 8 | 2612 | XP_016856059.1 | |
| AK091691.1 | 9 | 1717 | BAC03722.1 | |
| AK098092.1 | 11 | 2199 | BAC05230.1 | |
| AK295421.1 | 10 | 1886 | ||
| AY391836.1 | ||||
| AY461578.1 | 10 | 1905 | ||
| BC101373.1 | 10 | 1832 | AAI01374.1 | |
| BC101374.1 | 10 | 1832 | AAI01375.1 | |
| BC101375.1 | 10 | 1832 | AAI01376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv16e55 | Chr.1:241861796 - 242169657 on Build GRCh38 | Gain |
|
| nsv470788 | Chr.1:242037945 - 242392647 on Build GRCh38 | Gain |
|
| nsv528016 | Chr.1:242060097 - 242288048 on Build GRCh38 | Gain |
|
| nsv1002309 | Chr.1:241878940 - 242132333 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map