Assay Details
Target Gene Details
Entrez Gene ID: | 79902 |
Gene Name: | nucleoporin 85 |
Gene Aliases: |
FROUNT, Nup75 |
Location: |
Chr.17:75205450-75235759 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP85 | NM_001303276.1 | 1 | 83 | NP_001290205.1 |
| NM_024844.4 | 1 | 83 | NP_079120.1 | |
| XM_017025108.1 | 1 | 78 | XP_016880597.1 | |
| XM_017025109.1 | XP_016880598.1 | |||
| DA787201.1 | 1 | 97 | ||
| DC424627.1 | 1 | 85 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470602 | Chr.17:75090871 - 75208042 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map