Assay Details
Target Gene Details
Entrez Gene ID: | 51018 |
Gene Name: | ribosomal RNA processing 15 homolog |
Gene Aliases: |
CGI-115, KIAA0507 |
Location: |
Chr.1:218285270-218337983 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRP15 | NM_016052.3 | NP_057136.2 | ||
| XM_011509597.2 | XP_011507899.1 | |||
| AF151873.1 | AAD34110.1 | |||
| AK290066.1 | ||||
| BC020641.1 | AAH20641.1 | |||
| BF242786.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005829 | Chr.1:217815247 - 218302997 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map