Assay Details
Target Gene Details
Entrez Gene ID: | 51477 |
Gene Name: | inositol-3-phosphate synthase 1 |
Gene Aliases: |
INO1, INOS, IPS, IPS 1, IPS-1 |
Location: |
Chr.19:18434388-18438301 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISYNA1 | NM_001170938.1 | 9 | 1422 | NP_001164409.1 |
| NM_001253389.1 | 7 | 1288 | NP_001240318.1 | |
| NM_016368.4 | 10 | 1584 | NP_057452.1 | |
| NR_045573.1 | 9 | 1390 | ||
| NR_045574.1 | 7 | 1254 | ||
| XM_011528059.1 | 9 | 1663 | XP_011526361.1 | |
| AF086921.1 | 10 | 1403 | AAP97151.1 | |
| AF207640.1 | 10 | 1396 | AAG35698.1 | |
| AF220530.1 | 10 | 1416 | AAF26444.1 | |
| AK001325.1 | 9 | 1370 | BAA91626.1 | |
| AK021526.1 | 10 | 1416 | BAB13837.1 | |
| AK092179.1 | 9 | 1288 | ||
| AK092433.1 | 6 | 1503 | ||
| AK093461.1 | 7 | 1203 | ||
| AK296022.1 | 9 | 1252 | ||
| AL137749.1 | 7 | 1288 | CAB70904.1 | |
| BC004320.1 | 2 | 236 | AAH04320.1 | |
| BC018952.2 | 10 | 1399 | AAH18952.1 | |
| BC066902.1 | 10 | 1414 | AAH66902.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2677302 | Chr.19:18428535 - 18438882 on Build GRCh38 | Deletion |
|
| nsv952420 | Chr.19:18117791 - 18476990 on Build GRCh38 | Deletion |
|
| nsv482217 | Chr.19:18434815 - 18438133 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map