Assay Details
Target Gene Details
Entrez Gene ID: | 4714 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B8 |
Gene Aliases: |
ASHI, CI-ASHI |
Location: |
Chr.10:100523729-100529923 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFB8 | NM_001284367.1 | NP_001271296.1 | ||
| NM_001284368.1 | NP_001271297.1 | |||
| NM_005004.3 | NP_004995.1 | |||
| AA782554.1 | ||||
| AF044958.1 | AAD05422.1 | |||
| AF077028.1 | AAD27761.1 | |||
| AF115968.1 | AAP97239.1 | |||
| AK289579.1 | ||||
| AL080056.1 | CAB45691.1 | |||
| BC000466.2 | AAH00466.1 | |||
| BC019276.1 | AAH19276.1 | |||
| BM008944.1 | ||||
| BM686677.1 | ||||
| CF127298.1 | ||||
| CR533490.1 | CAG38521.1 | |||
| DA428446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624363 | Chr.10:100528312 - 100531647 on Build GRCh38 | Loss |
|
| nsv7519 | Chr.10:100517567 - 100539513 on Build GRCh38 | Insertion |
|
| nsv831962 | Chr.10:100406551 - 100593251 on Build GRCh38 | Gain |
|
| nsv831963 | Chr.10:100458231 - 100678922 on Build GRCh38 | Loss |
|
| esv2759779 | Chr.10:100392093 - 100805979 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map