Assay Details
Target Gene Details
Entrez Gene ID: | 10350 |
Gene Name: | ATP binding cassette subfamily A member 9 |
Gene Aliases: |
EST640918 |
Location: |
Chr.17:68974632-69078978 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCA9 | NM_080283.3 | NP_525022.2 | ||
| XM_017024011.1 | XP_016879500.1 | |||
| XM_017024012.1 | XP_016879501.1 | |||
| XM_017024013.1 | XP_016879502.1 | |||
| XM_017024014.1 | XP_016879503.1 | |||
| XM_017024015.1 | XP_016879504.1 | |||
| AF423307.1 | AAN32751.1 | |||
| AY028899.1 | AAK30024.1 | |||
| CR627382.1 | CAH10479.1 |
Target Gene Details
Entrez Gene ID: | 104355297 |
Gene Name: | ABCA9 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:69004714-69018323 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCA9-AS1 | NR_126414.1 | |||
| DA128020.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582891 | Chr.17:69006169 - 69012891 on Build GRCh38 | Loss |
|
| nsv833526 | Chr.17:68984617 - 69140893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map