Assay Details
Target Gene Details
Entrez Gene ID: | 64427 |
Gene Name: | tetratricopeptide repeat domain 31 |
Gene Aliases: |
- |
Location: |
Chr.2:74483032-74494564 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC31 | NM_022492.4 | 4 | 334 | NP_071937.4 |
| NR_027749.1 | 3 | 231 | ||
| XM_005264492.1 | 4 | 323 | XP_005264549.1 | |
| XM_011533035.1 | 4 | 323 | XP_011531337.1 | |
| XM_011533036.1 | 4 | 323 | XP_011531338.1 | |
| XM_011533037.1 | 4 | 323 | XP_011531339.1 | |
| XM_011533040.1 | 3 | 272 | XP_011531342.1 | |
| XM_017004706.1 | 4 | 323 | XP_016860195.1 | |
| XM_017004707.1 | 4 | 323 | XP_016860196.1 | |
| XM_017004708.1 | 3 | 228 | XP_016860197.1 | |
| XM_017004709.1 | 3 | 458 | XP_016860198.1 | |
| AK022850.1 | 4 | 320 | BAB14272.1 | |
| AK090520.1 | 3 | 373 | ||
| AK223355.1 | 4 | 318 | BAD97075.1 | |
| AK225626.1 | 4 | 320 | ||
| AK301585.1 | ||||
| AK303105.1 | 3 | 217 | ||
| AK303245.1 | ||||
| BC036017.1 | 4 | 318 | AAH36017.1 | |
| BC045536.1 | 3 | 222 | AAH45536.1 | |
| BC096709.1 | 4 | 320 | AAH96709.1 | |
| BC098365.1 | 4 | 320 | AAH98365.1 | |
| BX648131.1 | 3 | 216 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458185 | Chr.2:74367887 - 74531807 on Build GRCh38 | Loss |
|
| esv2759061 | Chr.2:74252797 - 74535298 on Build GRCh38 | Loss |
|

Genomic Map