Assay Details
Target Gene Details
Entrez Gene ID: | 84679 |
Gene Name: | solute carrier family 9 member A7 |
Gene Aliases: |
NHE-7, NHE7, SLC9A6 |
Location: |
Chr.X:46599251-46759172 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9A7 | NM_001257291.1 | 12 | 1695 | NP_001244220.1 |
| NM_032591.2 | 12 | 1692 | NP_115980.1 | |
| XM_005272681.4 | 12 | 1685 | XP_005272738.1 | |
| XM_005272682.4 | 12 | 1685 | XP_005272739.1 | |
| XM_006724564.3 | 12 | 1685 | XP_006724627.1 | |
| XM_011543990.2 | 12 | 1685 | XP_011542292.1 | |
| XM_017029905.1 | 12 | 1685 | XP_016885394.1 | |
| XM_017029906.1 | 12 | 1685 | XP_016885395.1 | |
| AF298591.1 | 12 | 1557 | AAK54508.1 | |
| AK056599.1 | 10 | 1020 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576853 | Chr.X:46169680 - 46644654 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map