Assay Details
Target Gene Details
Entrez Gene ID: | 28977 |
Gene Name: | mitochondrial ribosomal protein L42 |
Gene Aliases: |
HSPC204, L31MT, L42MT, MRP-L31, MRP-L42, MRP-S32, MRPL31, MRPS32, PTD007, RPML31, S32MT |
Location: |
Chr.12:93467490-93503772 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL42 | NM_014050.3 | NP_054769.1 | ||
| NM_172177.3 | NP_751917.1 | |||
| NR_038159.1 | ||||
| NR_038160.1 | ||||
| NR_038161.1 | ||||
| AF078860.1 | AAD44492.1 | |||
| AF135160.1 | AAF43784.1 | |||
| AF151038.1 | AAF36124.1 | |||
| AK000285.1 | BAA91054.1 | |||
| AL136659.1 | CAB66594.1 | |||
| BC040240.2 | AAH40240.1 | |||
| BM147739.1 | ||||
| BM726733.1 | ||||
| CD385500.1 | ||||
| CR533495.1 | CAG38526.1 | |||
| CR749344.1 | CAH18197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044791 | Chr.12:93470089 - 93489946 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map