Assay Details
Target Gene Details
Entrez Gene ID: | 353299 |
Gene Name: | regulator of G-protein signaling like 1 |
Gene Aliases: |
RGSL, RGSL2 |
Location: |
Chr.1:182447877-182560597 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map