Assay Details
Target Gene Details
Entrez Gene ID: | 116461 |
Gene Name: | tRNA splicing endonuclease subunit 15 |
Gene Aliases: |
C1orf19, PCH2F, sen15 |
Location: |
Chr.1:184050207-184095846 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSEN15 | NM_001127394.3 | 4 | 781 | NP_001120866.1 |
| NM_001300764.1 | 5 | 935 | NP_001287693.1 | |
| NM_001300766.1 | 5 | 819 | NP_001287695.1 | |
| NM_052965.3 | 5 | 923 | NP_443197.1 | |
| NR_023349.2 | 6 | 1040 | ||
| NR_125335.1 | 6 | 1003 | ||
| XM_011509139.2 | XP_011507441.1 | |||
| XM_017000228.1 | XP_016855717.1 | |||
| XM_017000229.1 | XP_016855718.1 | |||
| XM_017000230.1 | XP_016855719.1 | |||
| AF288394.1 | 5 | 898 | AAG60614.1 | |
| AK098196.1 | 5 | 821 | ||
| AK296655.1 | 4 | 676 | ||
| AK307241.1 | 6 | 931 | ||
| BC022030.1 | 5 | 868 | AAH22030.1 | |
| CB158796.1 | 1 | 165 |
More Information
Set Membership: |
|

Genomic Map