Assay Details
Target Gene Details
Entrez Gene ID: | 6296 |
Gene Name: | acyl-CoA synthetase medium-chain family member 3 |
Gene Aliases: |
SA, SAH |
Location: |
Chr.16:20763990-20797157 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSM3 | NM_005622.3 | 4 | 732 | NP_005613.2 |
| NM_202000.2 | 4 | 732 | NP_973729.1 | |
| XM_011545911.1 | 4 | 726 | XP_011544213.1 | |
| XM_017023523.1 | 4 | 726 | XP_016879012.1 | |
| XM_017023524.1 | 3 | 596 | XP_016879013.1 | |
| AK223139.1 | 4 | 701 | BAD96859.1 | |
| AK290322.1 | 4 | 701 | ||
| AK296122.1 | 3 | 420 | ||
| BC002790.2 | 3 | 591 | AAH02790.3 | |
| BI770558.1 | 4 | 732 | ||
| D16350.1 | 3 | 572 | BAA03853.1 | |
| X80062.1 | 4 | 681 | CAA56369.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050009 | Chr.16:20773077 - 20862801 on Build GRCh38 | Loss |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
| nsv833165 | Chr.16:20718218 - 20886396 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map