Assay Details
Target Gene Details
Entrez Gene ID: | 442117 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase-like 6 |
Gene Aliases: |
GALNACT20, GALNT17, GalNAc-T6L |
Location: |
Chr.4:171812690-173040408 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNTL6 | NM_001034845.2 | NP_001030017.2 | ||
| XM_017008243.1 | XP_016863732.1 | |||
| AK124240.1 | ||||
| BC047551.1 | AAH47551.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv596173 | Chr.4:171340402 - 172518114 on Build GRCh38 | Gain |
|
| nsv596171 | Chr.4:171184677 - 172359870 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map