Assay Details
Target Gene Details
Entrez Gene ID: | 51167 |
Gene Name: | cytochrome b5 reductase 4 |
Gene Aliases: |
NCB5OR, cb5/cb5R, dJ676J13.1 |
Location: |
Chr.6:83859651-83960427 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5R4 | NM_016230.3 | 2 | 252 | NP_057314.2 |
| AF169803.1 | AAF04812.1 | |||
| AK313146.1 | 2 | 225 | ||
| AL832113.1 | 2 | 217 | ||
| BC025380.1 | 2 | 219 | AAH25380.2 | |
| DA274558.1 | 2 | 257 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830718 | Chr.6:83781810 - 83941347 on Build GRCh38 | Gain+Loss |
|
| nsv1025798 | Chr.6:83654421 - 84334553 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map