Assay Details
Target Gene Details
Entrez Gene ID: | 2976 |
Gene Name: | general transcription factor IIIC subunit 2 |
Gene Aliases: |
TFIIIC-BETA, TFIIIC110 |
Location: |
Chr.2:27325849-27357034 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF3C2 | NM_001035521.2 | NP_001030598.1 | ||
| NM_001318909.1 | NP_001305838.1 | |||
| NM_001521.3 | NP_001512.1 | |||
| XM_005264272.4 | XP_005264329.1 | |||
| XM_011532801.2 | XP_011531103.1 | |||
| BC000212.2 | AAH00212.1 | |||
| BC020981.1 | AAH20981.1 | |||
| BP249089.1 | ||||
| BP278201.1 | ||||
| BT009799.1 | AAP88801.1 | |||
| BX647092.1 | ||||
| D13636.1 | BAA02800.1 | |||
| DB286393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833614 | Chr.2:27263559 - 27452213 on Build GRCh38 | Gain |
|
| esv3575209 | Chr.2:27334578 - 27389381 on Build GRCh38 | Gain |
|
| nsv581244 | Chr.2:27335385 - 27480628 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map