Assay Details
Target Gene Details
Entrez Gene ID: | 3383 |
Gene Name: | intercellular adhesion molecule 1 |
Gene Aliases: |
BB2, CD54, P3.58 |
Location: |
Chr.19:10270841-10286615 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ICAM1 | NM_000201.2 | 7 | 2814 | NP_000192.2 |
| AK130659.1 | 7 | 2535 | ||
| BC015969.2 | 7 | 2566 | AAH15969.1 | |
| J03132.1 | 7 | 2552 | AAA52709.1 | |
| M24283.1 | 7 | 2566 | AAA52708.1 | |
| M55100.1 | 1 | 435 |
More Information
Set Membership: |
|

Genomic Map