Assay Details
Target Gene Details
Entrez Gene ID: | 93974 |
Gene Name: | ATPase inhibitory factor 1 |
Gene Aliases: |
ATPI, ATPIP, IP |
Location: |
Chr.1:28236091-28238105 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATPIF1 | NM_016311.4 | NP_057395.1 | ||
| NM_178190.2 | NP_835497.1 | |||
| NM_178191.2 | 2 | 533 | NP_835498.1 | |
| AB029042.1 | BAA88422.1 | |||
| AK000934.1 | 2 | 470 | ||
| AK316600.1 | ||||
| AL050386.1 | 2 | 463 | CAI46227.1 | |
| AL583540.3 | ||||
| BC004955.1 | AAH04955.1 | |||
| BC009677.1 | AAH09677.1 | |||
| BG036978.1 | ||||
| BG337531.1 | ||||
| BG714388.1 | 2 | 526 | ||
| BP271519.1 | ||||
| BT009849.1 | AAP88851.1 | |||
| CR457097.1 | CAG33378.1 | |||
| DB030607.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545916 | Chr.1:28208722 - 28624350 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map