Assay Details
Target Gene Details
Entrez Gene ID: | 83879 |
Gene Name: | cell division cycle associated 7 |
Gene Aliases: |
ICF3, JPO1 |
Location: |
Chr.2:173354017-173368992 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDCA7 | NM_031942.4 | 10 | 2149 | NP_114148.3 |
| NM_145810.2 | 9 | 1912 | NP_665809.1 | |
| XM_011511957.2 | 10 | 2690 | XP_011510259.1 | |
| AK027628.1 | 10 | 2121 | BAB55245.1 | |
| AK027642.1 | 9 | 1893 | BAB55258.1 | |
| AK075134.1 | 8 | 2775 | BAC11425.1 | |
| AL833728.1 | 5 | 1441 | CAH56253.1 | |
| AL834186.1 | 7 | 2603 | CAH56357.1 | |
| AY029179.1 | 9 | 1925 | AAK31591.1 | |
| BC015124.1 | 5 | 1328 | AAH15124.1 | |
| BC027966.1 | 9 | 1878 | AAH27966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834462 | Chr.2:173327577 - 173506798 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78652913] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map