Assay Details
Target Gene Details
Entrez Gene ID: | 27123 |
Gene Name: | dickkopf WNT signaling pathway inhibitor 2 |
Gene Aliases: |
DKK-2 |
Location: |
Chr.4:106921802-107036296 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DKK2 | NM_014421.2 | 4 | 2919 | NP_055236.1 |
| AF177395.1 | 4 | 2931 | AAF02675.1 | |
| AY358414.1 | 4 | 2424 | AAQ88780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752020 | Chr.4:106720427 - 106966306 on Build GRCh38 | Gain |
|
| nsv4453 | Chr.4:106904437 - 106934973 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map