Assay Details
Target Gene Details
Entrez Gene ID: | 164832 |
Gene Name: | LON peptidase N-terminal domain and ring finger 2 |
Gene Aliases: |
RNF192 |
Location: |
Chr.2:100273291-100322733 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LONRF2 | NM_198461.3 | 12 | 6085 | NP_940863.3 |
| DQ020495.1 | 13 | 6037 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2675961 | Chr.2:100277996 - 100281375 on Build GRCh38 | Deletion |
|
| nsv834311 | Chr.2:100200455 - 100373574 on Build GRCh38 | Gain+Loss |
|
| esv3591751 | Chr.2:100278059 - 100281326 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map