Assay Details
Target Gene Details
Entrez Gene ID: | 146223 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 4 |
Gene Aliases: |
CKLFSF4 |
Location: |
Chr.16:66598178-66696707 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTM4 | NM_178818.2 | 5 | 2149 | NP_848933.1 |
| NM_181521.2 | 4 | 6852 | NP_852662.1 | |
| XM_011522882.1 | 4 | 6760 | XP_011521184.1 | |
| XM_017022954.1 | XP_016878443.1 | |||
| AF521889.1 | 5 | 2149 | AAN73845.1 | |
| AK000855.1 | ||||
| AK126493.1 | ||||
| AL157468.1 | ||||
| CR933620.1 | 1 | 5761 |
More Information
Additional Information:
For this assay, SNP(s) [rs144989651] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map