Assay Details
Target Gene Details
Entrez Gene ID: | 55967 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A12 |
Gene Aliases: |
B17.2, DAP13 |
Location: |
Chr.12:94971328-95003713 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA12 | NM_001258338.1 | 3 | 323 | NP_001245267.1 |
| NM_018838.4 | 4 | 411 | NP_061326.1 | |
| AF112208.1 | 4 | 431 | AAF17196.1 | |
| AF217092.1 | 4 | 390 | AAF91224.1 | |
| AK222619.1 | 4 | 398 | BAD96339.1 | |
| BC005936.1 | 4 | 400 | AAH05936.1 | |
| BG943528.1 | 4 | 396 | ||
| BT007220.1 | 4 | 378 | AAP35884.1 | |
| BU532909.1 | 3 | 310 | ||
| DW441120.1 | 3 | 337 |
More Information
Set Membership: |
|

Genomic Map