Assay Details
Target Gene Details
Entrez Gene ID: | 27068 |
Gene Name: | pyrophosphatase (inorganic) 2 |
Gene Aliases: |
HSPC124, SID6-306 |
Location: |
Chr.4:105369077-105474070 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPA2 | NM_006903.4 | 7 | 638 | NP_008834.3 |
| NM_176866.2 | 4 | 419 | NP_789842.2 | |
| NM_176867.3 | 2 | 227 | NP_789843.2 | |
| NM_176869.2 | 8 | 725 | NP_789845.1 | |
| AB026722.1 | 7 | 632 | BAA84701.1 | |
| AF161473.1 | 9 | 714 | AAF29088.1 | |
| AF217187.1 | 8 | 719 | AAG36781.1 | |
| AK000466.1 | 8 | 717 | BAA91184.1 | |
| AK289607.1 | 7 | 616 | ||
| AK299125.1 | 8 | 714 | ||
| AL136655.1 | 9 | 697 | CAB66590.2 | |
| AW953776.1 | ||||
| BC022803.1 | 2 | 203 | AAH22803.1 | |
| BC039462.1 | 4 | 366 | AAH39462.2 | |
| BC057219.1 | 7 | 631 | AAH57219.1 | |
| CN423093.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000196 | Chr.4:103475625 - 106148054 on Build GRCh38 | Loss |
|
| nsv470063 | Chr.4:104948391 - 105445052 on Build GRCh38 | Loss |
|
| nsv830027 | Chr.4:105300853 - 105485581 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map